missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cycloheximide, ultrapure, 95%

Catalog No. AAJ6690103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6690103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6690103 Supplier Thermo Scientific Chemicals Supplier No. J6690103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 66-81-9 | C15H23NO4 | 281.35 g/mol

A protein synthesis inhibitor in eukaryotic cells

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.35
MDL Number MFCD00082346
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
Synonym 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione
PubChem CID 6197
ChEBI CHEBI:27641
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1

Specifications

Type Cycloheximide
Physical Form Powder
Melting Point 111°C to 116°C
Color Cream to White
Quantity 1 g
Flash Point >110°C (230°F)
UN Number UN2811
Boiling Point 492°C
Beilstein 88868
Merck Index 14,2728
Solubility Information Soluble in DMSO, acetone, ethanol and methanol; Slightly soluble in water with sonication
Formula Weight 281.35
Percent Purity 95%
Chemical Name or Material Cycloheximide, Ultrapure
Show More Show Less
Hazard Category H300-H341-H360D
Hazard Statement GHS H Statement
H300-H360-H341
Fatal if swallowed.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-636-0
RTECSNumber MA4375000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only