missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cycloheximide, 95%

Catalog No. AC357420010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC357420010 1 g
AC357420050 5 g
AC357420250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC357420010 Supplier Thermo Scientific Chemicals Supplier No. 357420010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cycloheximide, CAS # 66-81-9, is a compound produced by the bacterium Streptomyces griseus that shows an antifungal antibiotic activity with the ability to inhibit protein synthesis only in eukaryotic cells.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cycloheximide is a semisynthetic compound showing an antifungal antibiotic activity.
  • It is produced by the bacterium Streptomyces griseus
  • Cycloheximide can inhibit protein synthesis in eukaryotic cells but not in prokaryotes by interacting directly with the translocase enzyme and interfering with the translocation step. Furthermore, this compound also blocks the A site of ribosomes

Application

  • Cycloheximide shows a neuroprotective, fungicide, and anticoronaviral activities
  • In biomedical research, it can induce glycogenolysis, gluconeogenesis, and ureagenesis
  • Due to significant toxic side effects such as teratogenesis, DNA damage, birth defects, and toxicity to sperm, this chemical compound is only used in in vitro research applications
  • This compound also shows inhibitory activity against ferroptosis
  • It is also used in laboratory media as a selective agent to permit the isolation of pathogenic and non-pathogenic fungi from other types of specimens
  • In laboratory research related to plant biology, it can induce ethylene production to promote the plant growth
  • In laboratory research, this compound can be applied to determine the half-life of protein in any cellular system. The treatment of in vitro cells with cycloheximide in a time-course experiment followed by western blotting using this cell lysate can show differences in protein half-life.

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.35
MDL Number MFCD00082346
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
Synonym cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide
PubChem CID 6197
ChEBI CHEBI:27641
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1

Specifications

Melting Point 107.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,2721
Specific Rotation Condition − 28.50 (20.00°C c=1,CHCl3)
Specific Rotation − 28.50
Solubility Information Solubility in water: 2.1 g/100mL (2 c). Other solubilities: soluble in chloroform, ether, acetone, methanol, and ethanol
Formula Weight 281.35
Percent Purity 95%
Physical Form Glassy Irregular Shaped Granules
Chemical Name or Material Cycloheximide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Suspected of causing genetic defects.
May damage the unborn child.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-636-

RUO – Research Use Only