Learn More
Cyclohexene oxide, 98%
CAS: 286-20-4 | C6H10O | 98.14 g/mol
Supplier: Thermo Scientific Chemicals 111215000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cyclohexene oxide | |
| 286-20-4 | |
| Colorless to Yellow | |
| 129.0°C to 130.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4520 to 1.4540 | |
| 500 mL | |
| 0.97 | |
| Solubility in water: insoluble in water. Other solubilities: soluble in alcohol, ether and acetone, soluble in most other organic solvents | |
| C1CCC2C(C1)O2 | |
| 98.14 | |
| 1.75 mPa.s (25°C) | |
| 98% |
| 98% | |
| -40.0°C | |
| 0.9700g/mL | |
| 27°C | |
| 97.5% min. (GC) | |
| C6H10O | |
| MFCD00005162 | |
| 17, 21 | |
| cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide | |
| ZWAJLVLEBYIOTI-UHFFFAOYSA-N | |
| 7-oxabicyclo[4.1.0]heptane | |
| 9246 | |
| 98.14 | |
| Liquid |
Chemical Identifiers
| 286-20-4 | |
| 98.14 | |
| ZWAJLVLEBYIOTI-UHFFFAOYSA-N | |
| 9246 | |
| C1CCC2C(C1)O2 |
| C6H10O | |
| MFCD00005162 | |
| cyclohexene oxide, 7-oxabicyclo 4.1.0 heptane, 1,2-epoxycyclohexane, cyclohexene epoxide, cyclohexylene oxide, tetramethyleneoxirane, cyclohexene 1-oxide, epoxycyclohexane, cyclohexeneoxide, 1,2-cyclohexene oxide | |
| 7-oxabicyclo[4.1.0]heptane |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 206-007-7
RTECSNumber : RN7175000
TSCA : TSCA
RUO – Research Use Only