missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene, 99% stab.

CAS: 110-83-8 | C6H10 | 82.146 g/mol

Supplier:  Thermo Scientific Chemicals A113590F

 View more versions of this product

Catalog No. AAA113590F


Only null left
Explore available promotions

Description

Description

Cyclohexene is used as a solvent. It is employed as a precursor to caprolactam, adipic acid, maleic acid, dicyclohexyladipate and cyclohexeneoxide. It is used as an intermediate for the production of cyclohexanol and cyclohexeneoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexene is used as a solvent. It is employed as a precursor to caprolactam, adipic acid, maleic acid, dicyclohexyladipate and cyclohexeneoxide. It is used as an intermediate for the production of cyclohexanol and cyclohexeneoxide.

Solubility
Immiscible with cold water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Cyclohexene
-104°C
83°C to 85°C
Petrol-like
1.446
2500 mL
906737
tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
HGCIXCUEYOPUTN-UHFFFAOYSA-N
cyclohexene
8079
82.15
110-83-8
0.811
−20°C (−4°F)
C6H10
MFCD00001539
UN2256
14,2727
Immiscible with cold water.
C1CCC=CC1
82.146
CHEBI:36404
99%

Chemical Identifiers

110-83-8
82.146
HGCIXCUEYOPUTN-UHFFFAOYSA-N
8079
cyclohexene
C6H10
MFCD00001539
tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
CHEBI:36404
C1CCC=CC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H302
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if swallowed.

P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P330-P331-P370+P378q-P501c

H225-H302-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEXENE

EINECSNumber : 203-807-8

RTECSNumber : GW2500000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only