missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene, 99% stab.

Catalog No. AAA11359AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11359AP 500 mL
AAA11359AE 100 mL
AAA113590F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA11359AP Supplier Thermo Scientific Chemicals Supplier No. A11359AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-83-8 | C6H10 | 82.146 g/mol

Cyclohexene is used as a solvent. It is employed as a precursor to caprolactam, adipic acid, maleic acid, dicyclohexyladipate and cyclohexeneoxide. It is used as an intermediate for the production of cyclohexanol and cyclohexeneoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexene is used as a solvent. It is employed as a precursor to caprolactam, adipic acid, maleic acid, dicyclohexyladipate and cyclohexeneoxide. It is used as an intermediate for the production of cyclohexanol and cyclohexeneoxide.

Solubility
Immiscible with cold water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 110-83-8
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00001539
InChI Key HGCIXCUEYOPUTN-UHFFFAOYSA-N
Synonym tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
PubChem CID 8079
ChEBI CHEBI:36404
IUPAC Name cyclohexene
SMILES C1CCC=CC1

Specifications

Melting Point -104°C
Density 0.811
Boiling Point 83°C to 85°C
Flash Point −20°C (−4°F)
Odor Petrol-like
Refractive Index 1.446
Quantity 500 mL
UN Number UN2256
Beilstein 906737
Merck Index 14,2727
Solubility Information Immiscible with cold water.
Formula Weight 82.15
Percent Purity 99%
Chemical Name or Material Cyclohexene
Show More Show Less
Hazard Category H225-H302-H304
Hazard Statement GHS H Statement
H225-H304-H302
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P330-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEXENE
EINECSNumber 203-807-8
RTECSNumber GW2500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only