missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexene, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 1 L |
| Packaging | Amber Glass Bottle |
| Percent Purity | 0.98 |
| Grade | Ungraded |
| Chemical Name or Material | Cyclohexene |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.