missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexene-1-carbonitrile 98.0+%, TCI America™
Supplier: TCI America C31911G
Specifications
| Cyclohexene-1-carbonitrile | |
| Yellow | |
| C7H9N | |
| 1 g | |
| 1-cyanocyclohexene, cyanocyclohexene, 1-cyclohexene-1-carbonitrile, cyclohexenecarbonitrile, 1-cyclohexenecarbonitrile, cyclohex-1-enecarbonitrile, cyclohex-1-ene-1-carbonitrile, 1-cyclohexenyl cyanide, 1-cyano-1-cyclohexene, acmc-1cqlm | |
| C1CCC(=CC1)C#N | |
| 107.156 | |
| 107.16 | |
| Liquid |
| 1855-63-6 | |
| 74°C | |
| MFCD00013772 | |
| 3276 | |
| GTMWGXABXQTZRJ-UHFFFAOYSA-N | |
| cyclohexene-1-carbonitrile | |
| 74619 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1855-63-6 | |
| 107.156 | |
| GTMWGXABXQTZRJ-UHFFFAOYSA-N | |
| 74619 | |
| C1CCC(=CC1)C#N |
| C7H9N | |
| MFCD00013772 | |
| 1-cyanocyclohexene, cyanocyclohexene, 1-cyclohexene-1-carbonitrile, cyclohexenecarbonitrile, 1-cyclohexenecarbonitrile, cyclohex-1-enecarbonitrile, cyclohex-1-ene-1-carbonitrile, 1-cyclohexenyl cyanide, 1-cyano-1-cyclohexene, acmc-1cqlm | |
| cyclohexene-1-carbonitrile |
Safety and Handling
TSCA : No