missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene-1-carbonitrile 98.0+%, TCI America™

Catalog No. C31915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31915G 5 g
C31911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31915G Supplier TCI America Supplier No. C31915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1855-63-6
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00013772
InChI Key GTMWGXABXQTZRJ-UHFFFAOYSA-N
Synonym 1-cyanocyclohexene, cyanocyclohexene, 1-cyclohexene-1-carbonitrile, cyclohexenecarbonitrile, 1-cyclohexenecarbonitrile, cyclohex-1-enecarbonitrile, cyclohex-1-ene-1-carbonitrile, 1-cyclohexenyl cyanide, 1-cyano-1-cyclohexene, acmc-1cqlm
PubChem CID 74619
IUPAC Name cyclohexene-1-carbonitrile
SMILES C1CCC(=CC1)C#N

Specifications

Color Yellow
Boiling Point 74°C
Quantity 5 g
UN Number 3276
Formula Weight 107.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexene-1-carbonitrile