missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, Reagents

Numéro de catalogue. 01337297
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01337297 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01337297 Fournisseur Labchem Inc Code fournisseur C2193600100C
Il en reste null

Cyclohexanone, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym Pimelic Ketone, Ketohexamethylene
IUPAC Name cyclohexanone
SMILES O=C1CCCCC1

Spécifications

Melting Point -28.9°C
Color Colorless or Pale Yellow
Quantity 100 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.9478
Boiling Point 154°C
Grade Reagent
Chemical Name or Material Cyclohexanone