missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanone, Reagents
Specifications
Specifications
| Melting Point | -28.9°C |
| Color | Colorless or Pale Yellow |
| Quantity | 1 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.9478 |
| Boiling Point | 154°C |
| Grade | Reagent |
| Chemical Name or Material | Cyclohexanone |