missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, ACS Reagent, Reagents

Catalog No. 01338026
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338026 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01338026 Supplier Labchem Inc Supplier No. C1193600100C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cyclohexanone, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym Pimelic Ketone, Ketohexamethylene
IUPAC Name cyclohexanone
SMILES O=C1CCCCC1

Specifications

Melting Point -28.9°C
Color Colorless or Pale Yellow
Quantity 100 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.9478
Boiling Point 154°C
Grade ACS
Chemical Name or Material Cyclohexanone, ACS Reagent