Learn More
Cyclohexanone, ACS, 99+%
CAS: 108-94-1 | C6H10O | 98.145 g/mol
Supplier: Thermo Scientific Chemicals 033309K7
Description
Cyclohexanone is used as an industrial solvent and paint remover. It acts as a precursor to nylon 6,6 and nylon 6 and cyclohexanone oxime, which gives caprolactam on rearrangement. Further, it is used as a chemical reaction medium, adhesives, sealants and agricultural products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| Cyclohexanone | |
| For ACS analysis | |
| Colorless | |
| 0.948 | |
| 4 L | |
| Liquid | |
| >99% | |
| MFCD00001625 | |
| 385735 | |
| Miscible with water and ethanol,acetone,ethyl ether and other organic solvents. | |
| C1CCC(=O)CC1 | |
| 98.145 | |
| CHEBI:17854 | |
| ACS |
| °C to 26°C | |
| 108-94-1 | |
| Colorless liquid | |
| 154°C to 156°C | |
| 47°C (116°F) | |
| 1.4507 | |
| C6H10O | |
| 4.5 hPa (3mm Hg) at 20°C | |
| ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon | |
| JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
| cyclohexanone | |
| 7967 | |
| 98.15 |
Identifiants chimiques
| 108-94-1 | |
| 98.145 | |
| JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
| 7967 | |
| cyclohexanone |
| C6H10O | |
| MFCD00001625 | |
| ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon | |
| CHEBI:17854 | |
| C1CCC(=O)CC1 |
Sécurité et manipulation
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H318-H335-H336
missing translation for 'dotInformation' : DOT Class: 3; Packing Group: III
missing translation for 'einecsNumber' : 203-631-1
missing translation for 'rtecsNumber' : GW1050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only