missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, ACS, 99+%

Catalog No. AA33309AP
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33309AP 500 mL
AA33309K2 1 L
AA33309M1 4 x 1 L
AA33309K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA33309AP Supplier Thermo Scientific Chemicals Supplier No. 033309AP

CAS: 108-94-1 | C6H10O | 98.145 g/mol

Cyclohexanone is used as an industrial solvent and paint remover. It acts as a precursor to nylon 6,6 and nylon 6 and cyclohexanone oxime, which gives caprolactam on rearrangement. Further, it is used as a chemical reaction medium, adhesives, sealants and agricultural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00001625
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1

Specifications

Melting Point °C to 26°C
Color Colorless
Appearance Colorless liquid
Density 0.947 g/cm3 at 20°C
Quantity 500 mL
Flash Point 47°C (116°F)
Physical Form Liquid
Refractive Index 1.4507
Assay Percent Range >99%
Vapor Pressure 4.5 hPa (3mm Hg) at 20°C
Beilstein 385735
Boiling Point 154°C to 156°C
Solubility Information Miscible with water and ethanol,acetone,ethyl ether and other organic solvents.
Formula Weight 98.15
Grade ACS
Chemical Name or Material Cyclohexanone, For ACS analysis
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H318-H335-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3; Packing Group: III
EINECSNumber 203-631-1
RTECSNumber GW1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only