missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, ACS, 99%, Spectrum™ Chemical

Catalog No. 18610105
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610105 100 mL Amber Glass Bottle
18610104 500 mL Amber Glass Bottle
18610106 1 L Amber Glass Bottle
18610107 4 L Amber Glass Bottle
18610102 20 L Steel Can
18610103 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610105 Supplier Spectrum Chemical Mfg Cor Supplier No. C1462100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 108-94-1

Specifications

CAS Max % 1
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.0005
Water 0.0005
Boiling Point 156°C
Percent Purity 0.99
Grade Reagent
Chemical Name or Material Cyclohexanone

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.