missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, 99+%

Catalog No. p-3345827
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15607AE 100 mL
AAA15607AP 500 mL
AAA156070F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA15607AE Supplier Thermo Scientific Chemicals Supplier No. A15607AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 108-94-1 | C6H10O | 98.145 g/mol

Cyclohexanone is used in the preparation of synthetic polymer such as nylon 6,6 and nylon 6. It acts as a solvent in organic synthesis. Its derivatives are used for the synthesis of dyes, rubber chemicals, plasticizers and pharmaceuticals. Further, it is involved in the preparation of cyclohexanone oxime, which rearranges to give caprolactam in the presence of sulfuric acid catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanone is used in the preparation of synthetic polymer such as nylon 6,6 and nylon 6. It acts as a solvent in organic synthesis. Its derivatives are used for the synthesis of dyes, rubber chemicals, plasticizers and pharmaceuticals. Further, it is involved in the preparation of cyclohexanone oxime, which rearranges to give caprolactam in the presence of sulfuric acid catalyst.

Solubility
Miscible with diethyl ether, acetone and alcohol. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, red metals, lead, amines, sulfuric acid, nitric acid, strong acids, aliphatic amines, rubber, resins and alkalies.

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00001625
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1

Specifications

Melting Point -26°C
Density 0.948
Quantity 100 mL
Flash Point 47°C (116°F)
Physical Form Liquid
Refractive Index 1.4507
Assay Percent Range ≥99%
UN Number UN1915
Beilstein 385735
Merck Index 14,2726
Boiling Point 154°C to 156°C
Solubility Information Miscible with diethyl ether,acetone and alcohol. Slightly miscible with water.
Formula Weight 98.15
Percent Purity ≥99%
Chemical Name or Material Cyclohexanone
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H318-H335-H336
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOHEXANONE
EINECSNumber 203-631-1
RTECSNumber GW1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only