missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanone, 99.0% min., ACS, MilliporeSigma™
Supplier: Technidata CX233520
Description
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Cyclohexanone | |
108-94-1 | |
Liquid | |
C6H10O | |
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon | |
C1CCC(=O)CC1 | |
98.145 | |
CHEBI:17854 | |
ACS |
For ACS analysis | |
200 L | |
99.0% min. | |
-CH2(CH2)4C(:O)- | |
JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
cyclohexanone | |
7967 | |
98.14 |
Chemical Identifiers
108-94-1 | |
98.145 | |
ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon | |
CHEBI:17854 | |
C1CCC(=O)CC1 |
C6H10O | |
JHIVVAPYMSGYDF-UHFFFAOYSA-N | |
7967 | |
cyclohexanone |
Safety and Handling
Recommended Storage : Store at +15°C to +25°C