missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Cyclohexanone

Catalog No. 398241500ML
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
398241500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 398241500ML Supplier Sigma Aldrich Supplier No. 398241500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cyclohexanone

Specifications

Melting Point -47°C
CAS 108-94-1
Density 0.947 g/mL at 25°C
Quantity 500 mL
Refractive Index n20/D 1.450
Molecular Formula C6H10O
Linear Formula C6H10(=O)
MDL Number MFCD00001625
Boiling Point 155°C
Molecular Weight (g/mol) 98.14 g/mol