missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanméthylamine 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 163°C |
| Quantity | 25 mL |
| UN Number | 2734 |
| Formula Weight | 113.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Cyclohexanemethylamine |
Sécurité et manipulation
| RTECSNumber | GV6128000 |
| TSCA | No |