Learn More
Cyclohexanemethylamine, 97%
CAS: 3218-02-8 | C4H6Br2O2 | 245.90 g/mol
$577.19 - $577.19
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC146570250
|
Thermo Scientific Chemicals
146570250 |
25 g | Glass bottle |
Each for $577.19
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3218-02-8 |
Specifications
| 3218-02-8 | |
| 0.8700g/mL | |
| 43°C | |
| 97.5% min. (GC) | |
| C4H6Br2O2 | |
| C6H11CH2NH2 | |
| 25 g | |
| 0.87 | |
| MELXIJRBKWTTJH-ONEGZZNKSA-N | |
| (E)-2,3-dibromobut-2-ene-1,4-diol | |
| 641240 | |
| 97% | |
| Cyclohexanemethylamine |
| Colorless to Yellow | |
| 161°C to 163°C | |
| Authentic | |
| Glass bottle | |
| 1.463 | |
| MFCD00004698 | |
| 12, 12 | |
| trans-2,3-dibromo-2-butene-1,4-diol, 2,3-dibromo-2-butene-1,4-diol, e-2,3-dibromobut-2-ene-1,4-diol, 2e-2,3-dibromobut-2-ene-1,4-diol, e-2,3-dibromo-2-butene-1,4-diol, 2,3-dibromobutene-1,4-diol, ccris 4781, 2-butene-1,4-diol, 2,3-dibromo-, 2e, 2-butene-1,4-diol, 2,3-dibromo, trans-2,3-dibromo-2-buten-1,4-diol | |
| OC\C(Br)=C(/Br)CO | |
| 245.90 | |
| 113.2 | |
| Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air
GHS Signal Word: Danger
EINECSNumber : 221-741-8
RUO – Research Use Only