missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. C047025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C047025G 25 g
C0470500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. C047025G missing translation for 'mfr' TCI America missing translation for 'supplierNo' C047025G

Chemical Identifiers

CAS 98-89-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00001461
InChI Key NZNMSOFKMUBTKW-UHFFFAOYSA-N
Synonym hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
PubChem CID 7413
ChEBI CHEBI:36096
IUPAC Name cyclohexanecarboxylic acid
SMILES C1CCC(CC1)C(=O)O

Specifications

Color White
Boiling Point 233°C
Quantity 25 g
Formula Weight 128.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Cyclohexanecarboxylic Acid
EINECSNumber (3)-3310
RTECSNumber GU8370000
TSCA Yes