missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarboxylic acid chloride, 98%

Catalog No. AC169051000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169051000 100 g Glass bottle
AC169050250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169051000 Supplier Thermo Scientific Chemicals Supplier No. 169051000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2719-27-9 | C7H11ClO | 146.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2719-27-9
Molecular Formula C7H11ClO
Molecular Weight (g/mol) 146.61
MDL Number MFCD00001456
InChI Key RVOJTCZRIKWHDX-UHFFFAOYSA-N
Synonym hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride
PubChem CID 75938
IUPAC Name cyclohexanecarbonyl chloride
SMILES ClC(=O)C1CCCCC1

Specifications

Color Colorless to Straw
Density 1.0900g/mL
Boiling Point 184°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4680 to 1.47
Linear Formula C6H11COCl
Quantity 100 g
Beilstein 09, 9
Specific Gravity 1.09
Solubility Information Solubility in water: hydrolyses in water. Other solubilities: soluble in toluene,methylene chloride,dioxane,,chlorobenzene,ethyl acetate,thf,dichloroethane
Formula Weight 146.62
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclohexanecarboxylic acid chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-322-7
TSCA TSCA

RUO – Research Use Only