missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarboxaldehyde 98.0+%, TCI America™

Catalog No. C088025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C088025ML 25 mL
C0880100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C088025ML Supplier TCI America Supplier No. C088025ML

Chemical Identifiers

CAS 2043-61-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001457
InChI Key KVFDZFBHBWTVID-UHFFFAOYSA-N
Synonym cyclohexanecarboxaldehyde, cyclohexanal, formylcyclohexane, cyclohexanealdehyde, cyclohexanaldehyde, cyclohexylcarboxaldehyde, 1-formylcyclohexane, hexahydrobenzaldehyde, cyclohexylformaldehyde, cyclohexane formaldehyde
PubChem CID 16275
IUPAC Name cyclohexanecarbaldehyde
SMILES O=CC1CCCCC1

Specifications

Color Colorless
Boiling Point 159°C
Quantity 25 mL
UN Number 1989
Formula Weight 112.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexanecarboxaldehyde
EINECSNumber (3)-3946
TSCA No