missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarbonyl chloride, 97+%

Catalog No. AAA1982422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1982422 100 g
AAA1982414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1982422 Supplier Thermo Scientific Chemicals Supplier No. A1982422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2719-27-9 | C7H11ClO | 146.61 g/mol

Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 2719-27-9
Molecular Formula C7H11ClO
Molecular Weight (g/mol) 146.61
MDL Number MFCD00001456
InChI Key RVOJTCZRIKWHDX-UHFFFAOYSA-N
Synonym hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride
PubChem CID 75938
IUPAC Name cyclohexanecarbonyl chloride
SMILES ClC(=O)C1CCCCC1

Specifications

Density 1.088
Boiling Point 183°C to 185°C
Flash Point 66°C (150°F)
Refractive Index 1.47
Quantity 100 g
UN Number UN3265
Beilstein 742163
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 146.62
Percent Purity ≥97%
Chemical Name or Material Cyclohexanecarbonyl chloride
Show More Show Less
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 220-322-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only