missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexaneacetic acid, 98%

CAS: 5292-21-7 | C8H14O2 | 142.20 g/mol

$81.92 - $303.63

Chemical Identifiers

CAS 5292-21-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001518
InChI Key LJOODBDWMQKMFB-UHFFFAOYSA-N
Synonym cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid
PubChem CID 21363
ChEBI CHEBI:37277
IUPAC Name 2-cyclohexylacetic acid
SMILES OC(=O)CC1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3198606
View Documents
Thermo Scientific Chemicals
H3198606
5 g
Each for $81.92
Only null left
 
AAH3198614
View Documents
Thermo Scientific Chemicals
H3198614
25 g
Each for $303.63
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5292-21-7
142.20
LJOODBDWMQKMFB-UHFFFAOYSA-N
21363
2-cyclohexylacetic acid
C8H14O2
MFCD00001518
cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid
CHEBI:37277
OC(=O)CC1CCCCC1

Specifications

5292-21-7
1.007
C8H14O2
5 g
cyclohexylacetic acid, cyclohexaneacetic acid, cyclohexylethanoic acid, hexahydrophenylacetic acid, cyclohexyl acetic acid, unii-v43k3to0hn, fema no. 2347, cyclohexane-acetic acid, v43k3to0hn, cyclo-hexylacetic acid
OC(=O)CC1CCCCC1
142.20
CHEBI:37277
98%
28°C to 32°C
242°C to 244°C
MFCD00001518
2041326
LJOODBDWMQKMFB-UHFFFAOYSA-N
2-cyclohexylacetic acid
21363
142.2
Cyclohexaneacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-132-0

RTECSNumber : GU6490000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only