missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane (Spectranalyzed™), Fisher Chemical™
For use in the ultraviolet and infrared ranges
$1361.17 - $1361.17
Chemical Identifiers
| CAS | 110-82-7 |
|---|---|
| Molecular Formula | C6H12 |
| Molecular Weight (g/mol) | 84.16 |
| MDL Number | MFCD00003814 |
| InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
| Synonym | hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane |
| PubChem CID | 8078 |
| ChEBI | CHEBI:29005 |
| IUPAC Name | cyclohexane |
| SMILES | C1CCCCC1 |
Chemical Identifiers
| 110-82-7 | |
| 84.16 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| 8078 | |
| cyclohexane |
| C6H12 | |
| MFCD00003814 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| CHEBI:29005 | |
| C1CCCCC1 |
Specifications
| 6.5°C | |
| Pass Test | |
| 0.770g/cm³ | |
| 4 L | |
| Pass Test | |
| Liquid | |
| ≥99.0 % | |
| CH2(CH2)4CH2 | |
| 104mbar at 20°C | |
| 1145 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| C1CCCCC1 | |
| 84.16 | |
| CHEBI:29005 | |
| ≥99.0% | |
| Cyclohexane |
| 110-82-7 | |
| Colorless | |
| 81°C | |
| Pass Test | |
| Amber Glass | |
| 0.002% max. | |
| C6H12 | |
| MFCD00003814 | |
| 0.94 mPa/s at 20°C | |
| 0.02% max. | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| cyclohexane | |
| 8078 | |
| 84.16 | |
| Also meets ACS specifications |
Safety and Handling
DOTInformation : DOT Class 3, : Flammable Liquid