missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, Puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120363
Change view
Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046188 1 L Glass Bottle
60046189 2.5 L Glass Bottle
60046194 6 x 1 L Glass Bottle
60046191 4 x 2.5 L Glass Bottle
60046190 30 kg Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60046188 Supplier Honeywell Chemicals Supplier No. 331171L
Only null left

Puriss. p.a., ACS Reagent,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 4°C to 7°C
Density 0.779g/mL at 25°C
Boiling Point 79°C to 81°C
Quantity 1 L
Explosion Limit 0.09
Flash Point −18°C (−.4°F)
Ignition Point 500°F
Packaging Glass Bottle
Refractive Index n20/D 1.4250 to 1.4280
Residue after Evaporation 0 to 0.001%
Vapor Pressure 168.8mmHg (37.7°C), 77mmHg (20°C)
UN Number UN1145
Beilstein 1900225
Formula Weight 84.16g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P261-P273-P301 + P310-P331-P501
Precautionary Statement H225-H304-H315-H336-H410
DOTInformation Class 3 - PG 2 - Cyclohexane
EINECSNumber 203-806-2
RTECSNumber GU6300000