missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, for pesticide residue analysis, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 326590010

Catalog No. AC326590010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For residue analysis
Specifications

Specifications

Cyclohexane
110-82-7
81.0°C
−18°C
1.4250 to 1.4270
0.77
C6H12
0.94 mPa.s (20°C)
05, 20
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
C1CCCCC1
84.16
CHEBI:29005
Pesticide
ECD Tested, For Pesticide Analysis
0.7700g/mL
1L
Glass bottle
2ppm max.
C6H12
MFCD00003814
0.02% max. (coulometric)
15, 2716
XDTMQSROBMDMFD-UHFFFAOYSA-N
cyclohexane
8078
84.15
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger

EINECSNumber : 203-806-2

RTECSNumber : GU6300000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only