missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, HPLC Grade, 99% min

Catalog No. p-4781884
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA22919K2 1 L
AA22919M1 4 x 1 L
AA22919K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA22919K2 Supplier Thermo Scientific Chemicals Supplier No. 022919K2
Only null left

CAS: 110-82-7 | C6H12 | 84.16 g/mol

Cyclohexane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Color Colorless
Appearance Colorless liquid
Density 0.779
Boiling Point 80°C to 81°C
Quantity 1 L
Flash Point -18°C (-1°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.426
Assay Percent Range 99% min.
Vapor Pressure 104 hPa (78mm Hg) at 20°C
Beilstein 1900225
Solubility Information Immiscible with water and soluble in ether,alcohol and acetone. Miscible with olive oil.
Formula Weight 84.16
Grade HPLC
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Category H225-H304-H315-H320-H336
Hazard Statement Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. Avoid contact with eyes. Take precautionary measures against static discharges. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 3; Packing Group: II
EINECSNumber 203-806-2
RTECSNumber GU6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only