missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane (HPLC), Fisher Chemical™

Catalog No. p-215865
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
Safe-Cote™ Amber Glass 
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C6201 1 L Amber Glass
C620SK1 1 L Safe-Cote™ Amber Glass 
C6204 4 L Amber Glass
C620SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. C6201 Supplier Thermo Fisher Scientific Supplier No. C6201
Only null left

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Absorbance 1.00 AU max. 202nm, 0.88 AU max. at 205nm, 0.67 AU max. at 210nm, 0.014 AU max. at 254nm
Color Colorless
Density 0.770g/cm³
Quantity 1 L
Packaging Amber Glass
Physical Form Liquid
Refractive Index 1.4235 to 1.4255 (at 25°C)
Residue after Evaporation 10ppm max.
Assay Percent Range 99.0% min by GC
Vapor Pressure 104mbar at 20°C
Linear Formula CH2(CH2)4CH2
Viscosity 0.94 mPa/s at 20°C
UN Number 1145
Water 0.02% max.
Boiling Point 81°C
Formula Weight 84.16
Percent Purity ≥99.0%
Grade Mobile phase for HPLC applications
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid