missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, For Spectroscopy Uvasol™, MilliporeSigma™

Catalog No. M1028222500
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1028222500 2.5 L Glass Bottle
M1028220500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1028222500 Supplier Technidata Supplier No. 1.02822.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6°C
Color Colorless
Density 0.78g/cm3 at 20°C
Quantity 2.5 L
Flash Point −18°C (−0.4°F)
Ignition Point 260°C (500°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.42
Vapor Pressure 103 hPa (20°C)
UN Number 1145
Boiling Point 81°C
Solubility Information Soluble in water: 55mg/L at 20°C
Formula Weight 84.16
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H304: May be fatal if swallowed and enters airways.
Precautionary Statement H315: Causes skin irritation.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.