missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, MilliporeSigma™
For environmental analysis
$735.58 - $735.58
Chemical Identifiers
| CAS | 110-82-7 |
|---|---|
| Molecular Formula | C6H12 |
| Molecular Weight (g/mol) | 84.16 |
| MDL Number | MFCD00003814 |
| InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
| Synonym | hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane |
| PubChem CID | 8078 |
| ChEBI | CHEBI:29005 |
| IUPAC Name | cyclohexane |
| SMILES | C1CCCCC1 |
Chemical Identifiers
| 110-82-7 | |
| 84.16 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| 8078 | |
| cyclohexane |
| C6H12 | |
| MFCD00003814 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| CHEBI:29005 | |
| C1CCCCC1 |
Specifications
| 6.7°C | |
| Colorless | |
| 80.6°C | |
| 1.2 to 8.3%(V) | |
| Pungent, fruity | |
| Liquid | |
| CH2(CH2)4CH2 | |
| 103 hPa (20°C) | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| cyclohexane | |
| 8078 | |
| 84.18 | |
| Cyclohexane |
| 110-82-7 | |
| 0.78g/cm3 at 20°C | |
| 4 L | |
| -20.15°C (-4.3°F) | |
| Glass Bottle | |
| C6H12 | |
| MFCD00003814 | |
| UN 1145 | |
| Insoluble in water | |
| C1CCCCC1 | |
| 84.16 | |
| CHEBI:29005 | |
| ≥99.0% |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging Group: II
Recommended Storage : Store at room temperature.