missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.5%, Honeywell Burdick & Jackson

Catalog No. p-7111733
Change view
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000483 6 x 1 L Glass Bottle
6000482 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000483 Supplier Honeywell Chemicals Supplier No. 0531L
Only null left

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Absorbance 0.17 AU max. at 225 nm (UV), 0.02 AU max. at 250 nm (UV), 0.005 AU max. at 300 nm (UV), 0.005 AU max. at 400 nm (UV)
CAS Max % 100.0000%
Density 0.78 g/cm3
Boiling Point 80.72°C
Quantity 6 x 1 L
Flash Point −20°C
Packaging Glass Bottle
UN Number UN1145
Beilstein 1900225
Water 0.01% max.
Solubility Information 0.6% in Water at 20°C
Formula Weight 84.16
Percent Purity >99.5%
Grade HPLC
Chemical Name or Material Cyclohexane, B&J Brand™, for HPLC, GC, pesticide residue analysis and spectrophotometry, >99.5%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Cyclohexane