missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, ACS reagent

Catalog No. p-4813430
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446280010 1 L Glass bottle
AC446280050 5 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446280010 Supplier Thermo Scientific Chemicals Supplier No. 446280010
Only null left

CAS: 110-82-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Density 0.7700g/mL
Boiling Point 81.0°C
Flash Point −18°C
Packaging Glass bottle
Linear Formula C6H12
Refractive Index 1.4250 to 1.4270
Quantity 1 L
Merck Index 15,2716
Specific Gravity 0.77
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
Viscosity 0.94 mPa.s (20°C)
Formula Weight 84.15
Grade ACS Reagent
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do
EINECSNumber 203-806-2

RUO – Research Use Only