missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, ACS Reagent, Reagents

Catalog No. p-200021041
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338029 1 L
01338028 20 L
01338027 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338029 Supplier Labchem Inc Supplier No. C11935001C
Only null left

Cyclohexane, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym Hexamethylene, Hexanaphthalene, Hexahydrobenzene
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Color Colorless
Boiling Point 80.7°C
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.7781
Grade ACS
Chemical Name or Material Cyclohexane, ACS Reagent