missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, 99+%

Catalog No. AAA16070AU
Change view
Click to view available options
Quantity:
1000 mL
5000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16070AU 1000 mL
AAA160700J 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16070AU Supplier Thermo Scientific Chemicals Supplier No. A16070AU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-82-7 | C6H12 | 84.16 g/mol

Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

Solubility
Miscible with ether, alcohol, acetone and olive oil. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Density 0.779
Quantity 1000 mL
Flash Point −18°C (−1°F)
Odor Pungent
Refractive Index 1.426
UN Number UN1145
Beilstein 1900225
Merck Index 14,2723
Boiling Point 80°C to 81°C
Solubility Information Miscible with ether,alcohol,acetone and olive oil. Immiscible with water.
Formula Weight 84.16
Percent Purity ≥99%
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Category H225-H304-H315-H320-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEXANE
EINECSNumber 203-806-2
RTECSNumber GU6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only