Learn More
Cyclohexane, 99+%, for spectroscopy
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Supplier: Thermo Scientific Chemicals 167740010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopySpecifications
| Cyclohexane | |
| 110-82-7 | |
| 0.0002 meq/g max. | |
| 1 L | |
| Liquid | |
| 99+% | |
| C6H12 | |
| Vapor Pressure: 104mbar at 20°C | |
| 05, 20 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| C1CCCCC1 | |
| 84.16 | |
| CHEBI:29005 | |
| ≥99.0% |
| 99+% | |
| 0.01 max. at 260nm, 0.02 max. at 250nm, 0.09 max. at 235nm, 0.3 max. at 220nm, 1 max. at 210nm | |
| 2.90 (Air = 1.0) | |
| Glass Bottle | |
| 0.0005% max. | |
| C6H12 | |
| MFCD00003814 | |
| 0.01% max. | |
| 15, 2716 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| cyclohexane | |
| 8078 | |
| 84.15 | |
| Spectroscopy |
Chemical Identifiers
| 110-82-7 | |
| 84.16 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| 8078 | |
| cyclohexane |
| C6H12 | |
| MFCD00003814 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| CHEBI:29005 | |
| C1CCCCC1 |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 203-806-2
RTECSNumber : GU6300000
TSCA : TSCA
RUO – Research Use Only