Learn More
Cyclohexane, 99+%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur.
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Supplier: Thermo Scientific Chemicals 406020025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopySpecifications
| Cyclohexane | |
| 110-82-7 | |
| 0.7700g/mL | |
| 2.5 L | |
| Glass Bottle | |
| 1.4250 to 1.4270 | |
| 99+% | |
| C6H12 | |
| MFCD00003814 | |
| 0.02% max. (Coulometric) | |
| 15, 2716 | |
| Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4. | |
| C1CCCCC1 | |
| 84.16 | |
| CHEBI:29005 | |
| ≥99% |
| 6.5°C | |
| 0.01 max. at 300nm, 0.01 max. at 400nm, 0.02 max. at 260nm, 0.03 max. at 250nm, 0.08 max. at 240nm, 0.2 max. at 230nm, 0.5 max. at 220nm, 1 max. at 210nm | |
| 81.0°C | |
| −18°C | |
| Liquid | |
| 0.002% max. | |
| 0.77 | |
| C6H12 | |
| 0.94 mPa.s (20°C) | |
| 05, 20 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| cyclohexane | |
| 8078 | |
| 84.15 | |
| ACS |
Chemical Identifiers
| 110-82-7 | |
| 84.16 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| 8078 | |
| cyclohexane |
| C6H12 | |
| MFCD00003814 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| CHEBI:29005 | |
| C1CCCCC1 |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Danger
EINECSNumber : 203-806-2
RTECSNumber : GU6300000
TSCA : TSCA
RUO – Research Use Only