missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, 99+%, for spectroscopy ACS, meets the requirements of Reag.Ph.Eur.

Catalog No. AC406025000
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406025000 500 mL Glass Bottle
AC406020025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC406025000 Supplier Thermo Scientific Chemicals Supplier No. 406025000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110-82-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Absorbance 0.01 max. at 300nm, 0.01 max. at 400nm, 0.02 max. at 260nm, 0.03 max. at 250nm, 0.08 max. at 240nm, 0.2 max. at 230nm, 0.5 max. at 220nm, 1 max. at 210nm
Density 0.7700g/mL
Quantity 500 mL
Flash Point −18°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4250 to 1.4270
Residue after Evaporation 0.002% max.
Specific Gravity 0.77
Assay Percent Range 99+%
Linear Formula C6H12
Viscosity 0.94 mPa.s (20°C)
Water 0.02% max. (Coulometric)
Beilstein 05, 20
Merck Index 15, 2716
Boiling Point 81.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
Formula Weight 84.15
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Cyclohexane
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 203-806-2
RTECSNumber GU6300000
TSCA TSCA

RUO – Research Use Only