missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, 99.8%, for HPLC

Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
Specifications
Specifications
| Melting Point | 6.5°C |
| Absorbance | (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.004 max. at 240nm, 0.004 max. at 250nm, 0.018 max. at 230nm, 0.097 max. at 220nm, 0.201 max. at 210nm, 0.301 max. at 200nm |
| Color | White |
| Density | 0.7700g/mL |
| Quantity | 1 L |
| Flash Point | -18°C |
| Packaging | Glass Bottle |
| Physical Form | Crystalline Solid |
| Refractive Index | 1.4250 to 1.4270 |
| Residue after Evaporation | 0.0002 % max. |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. |
| EINECSNumber | 203-806-2 |
| RTECSNumber | GU6300000 |
| TSCA | TSCA |
RUO – Research Use Only