missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, 99.5%, Extra Dry, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 6.5°C |
| Acidity | 0.001 meq/g max. |
| Density | 0.7700g/mL |
| Boiling Point | 81.0°C |
| Quantity | 100 mL |
| Flash Point | −18°C |
| Infrared Spectrum | Authentic |
| Packaging | AcroSeal™ Glass Bottle |
| Physical Form | Liquid |
| Refractive Index | 1.4250 to 1.4270 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | Danger |
| missing translation for 'healthHazard2' | GHS H Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. |
| missing translation for 'healthHazard3' | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. |
| EINECSNumber | 203-806-2 |
| RTECSNumber | GU6300000 |
| TSCA | TSCA |
RUO – Research Use Only