missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, 99.0% min., ACS, MilliporeSigma™
Supplier: Technidata CX22903
Specifications
| Cyclohexane | |
| 110-82-7 | |
| Liquid | |
| C6H12 | |
| MFCD00003814 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| cyclohexane | |
| 8078 | |
| 84.16 |
| For ACS analysis | |
| 4 L | |
| 99.0%, min. | |
| -CH2(CH2)4CH2- | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| C1CCCCC1 | |
| 84.16 | |
| CHEBI:29005 | |
| ACS/NF/USP |
Chemical Identifiers
| 110-82-7 | |
| 84.16 | |
| XDTMQSROBMDMFD-UHFFFAOYSA-N | |
| 8078 | |
| cyclohexane |
| C6H12 | |
| MFCD00003814 | |
| hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
| CHEBI:29005 | |
| C1CCCCC1 |