missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptylamine 97.0+%, TCI America™

Catalog No. C067910G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C067910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C067910G Supplier TCI America Supplier No. C067910G
Only null left

Chemical Identifiers

CAS 5452-35-7
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.204
MDL Number MFCD00004153
InChI Key VXVVUHQULXCUPF-UHFFFAOYSA-N
Synonym cycloheptylamine, aminocycloheptane, cycloheptyl amine, cycloheptanamine, n-cycloheptylamine, 1-cycloheptylamine, cycloheptanyl amine, sz1, acmc-209lh8
PubChem CID 2899
IUPAC Name cycloheptanamine
SMILES C1CCCC(CC1)N

Specifications

Color Yellow
Boiling Point 170°C
Quantity 10 g
UN Number 2733
Formula Weight 113.20
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cycloheptylamine