missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptylacetic acid, 99%

Catalog No. AAL1555303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1555303 1 g
AAL1555306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1555303 Supplier Thermo Scientific Chemicals Supplier No. L1555303
Only null left

CAS: 4401-20-1 | C9H16O2 | 156.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4401-20-1
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD01075701
InChI Key DQNWKASPZFJVMJ-UHFFFAOYSA-N
Synonym cycloheptylacetic acid, cycloheptaneacetic acid, 2-cycloheptylaceticacid, pubchem16200, acmc-20ald3
PubChem CID 227401
IUPAC Name 2-cycloheptylacetic acid
SMILES C1CCCC(CC1)CC(=O)O

Specifications

Melting Point 14°C to 15°C
Density 1.023
Boiling Point 110°C to 112°C (1 mmHg)
Refractive Index 1.4725
Quantity 1 g
Beilstein 1935484
Formula Weight 156.23
Percent Purity 99%
Chemical Name or Material Cycloheptylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No

RUO – Research Use Only