missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptyl bromide, 97%, Thermo Scientific™

Catalog No. p-96517
Change view
Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111080250 25g Glass bottle
AC111081000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111080250 Supplier Thermo Scientific Chemicals Supplier No. 111080250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 2404-35-5
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD00004149
InChI Key LOXORFRCPXUORP-UHFFFAOYSA-N
Synonym cycloheptane, bromo, cycloheptyl bromide, cycloheptylbromide, bromocyclo-heptane, 2-bromocycloheptane, bromocycloheptane, acmc-209g8c
PubChem CID 16992
IUPAC Name bromocycloheptane
SMILES BrC1CCCCCC1

Specifications

Density 1.2800g/mL
Boiling Point 71°C to 72°C (10.0mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5035 to 1.5055
Linear Formula C7H13Br
Quantity 25g
Beilstein 05, 29
Specific Gravity 1.28
Formula Weight 177.08
Percent Purity 97%
Chemical Name or Material Cycloheptyl bromide, 97%
Show More Show Less
EINECSNumber 219-294-9
TSCA TSCA

RUO – Research Use Only