missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptene 96.0+%, TCI America™

Catalog No. p-7124563
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06885ML 5 mL
C068825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06885ML Supplier TCI America Supplier No. C06885ML
Only null left

Chemical Identifiers

CAS 628-92-2
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00004156
InChI Key ZXIJMRYMVAMXQP-UHFFFAOYSA-N
Synonym cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure
PubChem CID 12363
IUPAC Name cycloheptene
SMILES C1CCC=CCC1

Specifications

Color Colorless
Boiling Point 115°C
Quantity 5 mL
UN Number 2242
Formula Weight 96.17
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Cycloheptene