missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptene, 96%, stab. with 0.1% BHT

Catalog No. AAH2906206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2906206 5 g
AAH2906214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2906206 Supplier Thermo Scientific Chemicals Supplier No. H2906206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 628-92-2 | C7H12 | 96.173 g/mol

Cycloheptene was used in one-pot synthesis of 1,2/3-triols from the allylichydroperoxides catalyzed by zeolite-confined osmium(0) nanoclusters. It is a raw material in organic chemistry and a monomer in polymer synthesis. Copper (1) triflate catalyzes the photodimerization of the simple nonconjugated unstrained olefins, cyclopentene, cyclohexene, and cycloheptene, but not cyclooctene or acyclic olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cycloheptene was used in one-pot synthesis of 1,2/3-triols from the allylichydroperoxides catalyzed by zeolite-confined osmium(0) nanoclusters. It is a raw material in organic chemistry and a monomer in polymer synthesis. Copper (1) triflate catalyzes the photodimerization of the simple nonconjugated unstrained olefins, cyclopentene, cyclohexene, and cycloheptene, but not cyclooctene or acyclic olefins.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids. May react exothermically with reducing agents to release hydrogen gas.

Chemical Identifiers

CAS 628-92-2
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00004156
InChI Key ZXIJMRYMVAMXQP-UHFFFAOYSA-N
Synonym cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure
PubChem CID 12363
IUPAC Name cycloheptene
SMILES C1CCC=CCC1

Specifications

Density 0.833
Boiling Point 112°C to 115°C
Flash Point −7°C (19°F)
Refractive Index 1.458
Quantity 5 g
UN Number UN2242
Solubility Information Insoluble in water.
Formula Weight 96.17
Percent Purity 96%
Chemical Name or Material Cycloheptene, stabilized with 0.1% BHT
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOHEPTENE
EINECSNumber 211-060-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only