missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptene, 95%, stabilized

Catalog No. AC304260250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC304260250 25 g
AC304260050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC304260250 Supplier Thermo Scientific Chemicals Supplier No. 304260250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 628-92-2 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 628-92-2
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00004156
InChI Key ZXIJMRYMVAMXQP-UHFFFAOYSA-N
Synonym cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure
PubChem CID 12363
IUPAC Name cycloheptene
SMILES C1CCC=CCC1

Specifications

Melting Point -56°C
Color Colorless
Density 0.8200g/mL
Boiling Point 114.6°C
Flash Point -006°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4570 to 1.459
Linear Formula C7H12
Quantity 25 g
Beilstein 05, 65
Specific Gravity 0.82
Solubility Information Solubility in water: immiscible with water. Other solubilities: soluble in ether,ethanol,benzene
Formula Weight 96.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Cycloheptene, 90%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-060-4

RUO – Research Use Only