missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanone 98.0+%, TCI America™

Catalog No. C0466500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0466500ML 500 mL
C046625ML 25 mL
C0466100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0466500ML Supplier TCI America Supplier No. C0466500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 502-42-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00004159
InChI Key CGZZMOTZOONQIA-UHFFFAOYSA-N
Synonym suberone, ketocycloheptane, suberon, ketoheptamethylene, cycloheptanon, cycloheptyloxy, chembl18607, cycloheptan-1-one, cyclo-heptanone, 1-oxocycloheptane
PubChem CID 10400
IUPAC Name cycloheptanone
SMILES C1CCCC(=O)CC1

Specifications

Melting Point -21°C
Color Yellow
Boiling Point 180°C
Quantity 500 mL
UN Number 1224
Formula Weight 112.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cycloheptanone
RTECSNumber GU3325000
TSCA Yes