missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanone, 98+%

Catalog No. p-7021189
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1342118 50 g
AAA1342130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1342118 Supplier Thermo Scientific Chemicals Supplier No. A1342118

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 502-42-1 | C7H12O | 112.172 g/mol

Cycloheptanone is used as a precursor for the synthesis of pharmaceuticals. A spasmolytic agent and vasodilator is produced from this chemical. They are also used in certain stereospecific enzymatic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cycloheptanone is used as a precursor for the synthesis of pharmaceuticals. A spasmolytic agent and vasodilator is produced from this chemical. They are also used in certain stereospecific enzymatic reactions.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents,strong bases, reducing agents.

Chemical Identifiers

CAS 502-42-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00004159
InChI Key CGZZMOTZOONQIA-UHFFFAOYSA-N
Synonym suberone, ketocycloheptane, suberon, ketoheptamethylene, cycloheptanon, cycloheptyloxy, chembl18607, cycloheptan-1-one, cyclo-heptanone, 1-oxocycloheptane
PubChem CID 10400
IUPAC Name cycloheptanone
SMILES C1CCCC(=O)CC1

Specifications

Melting Point -21°C
Density 0.956
Boiling Point 179°C to 181°C
Flash Point 55°C (131°F)
Odor Sweet
Refractive Index 1.461
Quantity 50 g
UN Number UN1224
Beilstein 969823
Merck Index 14,2722
Solubility Information It is insoluble in water.
Formula Weight 112.17
Percent Purity 99%
Chemical Name or Material Cycloheptanone
Show More Show Less
Hazard Category H226-H302+H332-H318
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 207-937-6
RTECSNumber GU3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only