missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptanol 98.0+%, TCI America™

Catalog No. C067825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C067825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C067825ML Supplier TCI America Supplier No. C067825ML
Only null left

Chemical Identifiers

CAS 502-41-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00004150
InChI Key QCRFMSUKWRQZEM-UHFFFAOYSA-N
Synonym suberol, hydroxycycloheptane, 1-cycloheptanol, suberyl alcohol, cycloheptanol, acmc-209kko
PubChem CID 10399
IUPAC Name cycloheptanol
SMILES C1CCCC(CC1)O

Specifications

Color Yellow
Boiling Point 185°C
Quantity 25 mL
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cycloheptanol